missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ 60mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 60mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
£369.00 - £790.00
Specifications
Capacity (Metric) | 125 mL |
---|---|
Diameter (Metric) Stem | 13mm |
Material | Pyrex Glass, Borosilicate Glass |
Height (Metric) | 180 mm |
For Use With (Application) | For convenient filtration of solutions |
Product Code | Brand | Porosity | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Pore Size | Price | Quantity & Availability | ||||
12613222
|
Pyrex
3700/02 |
1 | 100 to 160μm |
£395.00
Pack of 5 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
11317994
|
Pyrex
3700/04 |
2 | 40 to 100μm |
£395.00
Pack of 5 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
11900955
|
Pyrex
3700/01 |
0 | 160 to 250μm |
£369.00
Pack of 5 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
11981025
|
Pyrex
3700/06 |
3 | 16 to 40μm |
£374.00
Pack of 5 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
12643222
|
Pyrex
3700/08 |
4 | 10 to 16μm |
£373.00
Pack of 5 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
15567710
|
Pyrex
3700/10 |
5 | 1.0 to 1.6μm |
£790.00
Pack of 10 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||||
Description
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 60mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
- Choice of disc diameters and porosities
Specifications
125 mL | |
Pyrex Glass, Borosilicate Glass | |
For convenient filtration of solutions | |
Integrated filter plate sintered Glass disc |
13mm | |
180 mm | |
60mm | |
100mm |