missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Punt Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£203.00 - £559.00
Specifications
| Antigen | Punt |
|---|---|
| Concentration | 0.75 mg/mL |
| Dilution | Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18059363
|
Novus Biologicals
NB300-662-0.025MG |
0.025 mg |
£203.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18229124
|
Novus Biologicals
NB300-662-0.05MG |
0.05 mg |
£341.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18296520
|
Novus Biologicals
NB300-662 |
0.1 mg |
£559.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Punt Polyclonal specifically detects Punt in Drosophila samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Punt | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Activin A receptor 88CD, Activin receptor, Activin receptor type II, ATR, Atr II, Atr88CD, AtrII, CG7904, dlhC, Dorsal holes C, j5A5, l(3)10460, Pun, Punt receptor serine/threonine kinase, Put, STK C, STKC, TGF B, TGF beta, TGF beta l, TGF beta type receptor like, Tgf r, TGFB, TGFbeta | |
| IgG | |
| Punt |
| 0.75 mg/mL | |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Drosophila | |
| Synthetic peptide: KSTTEATTQVPKEKTQDGSN[C]. Peptides was amidated for conjugation. | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title