missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Punt Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-662-0.05MG
This item is not returnable.
View return policy
Description
Punt Polyclonal specifically detects Punt in Drosophila samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Punt | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide | |
| Rabbit | |
| 0.05 mg | |
| Punt | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.75 mg/mL | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500 | |
| Activin A receptor 88CD, Activin receptor, Activin receptor type II, ATR, Atr II, Atr88CD, AtrII, CG7904, dlhC, Dorsal holes C, j5A5, l(3)10460, Pun, Punt receptor serine/threonine kinase, Put, STK C, STKC, TGF B, TGF beta, TGF beta l, TGF beta type receptor like, Tgf r, TGFB, TGFbeta | |
| Synthetic peptide: KSTTEATTQVPKEKTQDGSN[C]. Peptides was amidated for conjugation. | |
| Primary | |
| Drosophila | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction