missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Punt Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-662
This item is not returnable.
View return policy
Description
Punt Polyclonal specifically detects Punt in Drosophila samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Punt | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Drosophila | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.75 mg/mL | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500 | |
| Activin A receptor 88CD, Activin receptor, Activin receptor type II, ATR, Atr II, Atr88CD, AtrII, CG7904, dlhC, Dorsal holes C, j5A5, l(3)10460, Pun, Punt receptor serine/threonine kinase, Put, STK C, STKC, TGF B, TGF beta, TGF beta l, TGF beta type receptor like, Tgf r, TGFB, TGFbeta | |
| Synthetic peptide: KSTTEATTQVPKEKTQDGSN[C]. Peptides was amidated for conjugation. | |
| 0.1 mg | |
| Signal Transduction | |
| Punt | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction