missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Beaker with Spout

Product Code. 10044630 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
1 mL
5 mL
10 mL
25 mL
50 mL
100 mL
150 mL
250 mL
400 mL
500 mL
600 mL
1000 mL
2000 mL
5000 mL
Unit Size:
Each
Pack of 5
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10044630 1000 mL Each
16236511 1 mL Pack of 5
16246511 5 mL Pack of 5
16256511 10 mL Pack of 5
10288380 25 mL Each
10628461 50 mL Each
10792921 100 mL Each
10248480 150 mL Each
10713101 250 mL Each
10094800 400 mL Each
10349740 500 mL Each
10257810 600 mL Each
10762751 2000 mL Each
10237860 5000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10044630
Only null left
Add to basket
Add to basket

Includes a machined base for good heat transfer. The PTFE beaker includes a molded-in spout for easy pouring.

Non-contaminating and inert to highly corrosive reagents.
  • Non-wettable, lightweight, and heat resistant up to 280°C
  • Machined base for good heat transfer
  • Easy to clean
  • Molded-in spout for easy pouring
  • Isostatically molded from pure PTFE

Specifications

Color White
Diameter (Metric) Outer 100mm
Diameter (Metric)Top 100mm
Capacity (Metric) 1000 mL
Material PTFE
Height (Metric) 155mm
Temperature (Metric) Max. Operating 280°C
Type Beaker
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.