Learn More
RETSCH PT 200 Model Rotating Tube Dividers
Brand:  RETSCH 40.412.0004
Code : NEW
Additional Details : Weight : 37.00000kg
 View more versions of this product
Product Code. 11914202
  • £6175.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket
Specifications
Rotating Tube Divider
100/240 V, 50/60 Hz
1050 mm
551 mm
1-phase Power Connection
∼46 kg
551 x 520 x 1050 mm
Agriculture, biology, chemistry/Plastics, construction materials, engineering/electronics, environment/recycling, food, geology/metallurgy, glass/ceramics, medicine/pharmaceuticals
50/60 Hz
PT 200
100/240 V
520 mm