missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For Research Use Only. Not for use in diagnostic procedures.

Brand:  Agilent BioTek N2SA-SN


Additional Details : Weight : 43.00000kg

 View more versions of this product

Product Code. 17248039

  • POA / Each
Estimated Shipment: 20-08-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. Variable bandwidth quad monochromators, sensitive high transmission filter-based optics, laser TRF, and up to 4 PMTs provide ultrafast measurements.

Advanced environmental controls, including CO2/O2 control, incubation to 70°C, and variable shaking support live cell assays. BioStack Neo delivers walk-away automation, high throughput, barcode-labeled filter cubes to streamline workflows and limit errors. Powerful Gen5 software provides complete reader control, powerful data analysis, and LIMS integration.

Specifications

Specifications

230 to 999 nm, 1 nm increments
CE and TUV marked. RoHS compliant.
52.5 cm
±0.2°C at 37°C
0 to 4 OD
HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP
41 cm
Monochromater: ABS; Filter: Single filter top FI/FP/TRF and LUM, Alpha Laser
Multi-Mode Microplate Reader
0.0001 OD
35 kg
39 cm
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD
0 to 20%
20.7 in.
Linear, Orbital, Double Orbital
5 to 1000 μL, 1 μL increments
16.1 in.
≤1% from 0 to 3 OD
NEO2SALPHA
250 W max.
78 lb.
15.4 in.
Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option)
Xenon Flash
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers