Learn More
Agilent BioTek Peltier Cooling Module
Peltier Cooling Module helps maintain internal environmental stability and promotes rapid cool-down.
Brand:  Agilent BioTek 1700001-S
Additional Details : Weight : 6.80000kg
Product Code. 16542172
  • £11700.00 / Each
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
  • The Peltier Cooling Module promotes a rapid interior cool down after incubated processes, to allow efficient switching between multiple applications without unwanted temperature influences
  • For temperature sensitive assays typically run at ambient, or room temperature, the Cooling Module maintains environmental stability within Cytation, allowing 1 degree celcius rise in ambient temperature, regardless of external and internal temperature fluctuation
Specifications
Specifications
Cytation 5 and Cytation 1
ELISA, Fluorescence ELISA, Protein Quantification, Nucleic Acid Quantification