Learn More
WTW™ Multiparameter Benchtop Meter InoLab™ Multi 9630 IDS
The WTW™ Multiparameter Benchtop Meter InoLab™ Multi 9630 IDS Digital high performance multi-parameter device with three inputs for pH, ISE, conductivity and dissolved oxygen, with comprehensive GLP functions.
Brand:  WTW™ 1FD57K
Additional Details : Weight : 2.50000kg
 View more versions of this product
Product Code. 15582016
  • £4225.00 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
  • Modern high-performance multi-parameter meter with three galvanically isolated measuring channels for pH/mV, ISE (including incremental process), conductivity and oxygen measurement
  • Up to three of the same or different parameters can be measured
  • Fatigue-proof reading on brilliant large 16:9 color graphic display under a hygienic glass shield and antibacterial keyboard for use even in critical areas
  • For use with wireless modules together and IDS plug head sensors for independence from cables, permitting convenient use under laboratory hoods or laminar flow benches
  • Solid zinc die casting lower housing adds safety against interference and ensures stability on the lab bench
  • Two USB interfaces communicate with a computer, printer or USB memory stick allow the transfer of all data
  • Activatable user administration for automatic GLP documentation ensures additional safety and traceability
  • Integratable adapter for conventional pH/ORP/ISE electrodes available
  • Includes MultiLab™ Importer software to capture data via Excel™ as well as via MultiLab™ User (user administration)
Specifications
Specifications
USB A/Mini USB-B
3
pH, Temperature, mV, Saturation, Saturation, Concentration, Conductivity, spec. resistance, spec. resistance, TDS, TDS
Inolab™ Multi 9630 IDS, Set K
Depends on the IDS sensor used
LCD
Meter, including pH electrode SenTix™ 980, IDS oxygen sensor FDO™ 925, IDS conductivity cell TetraCon™ 925
Universal power supply 100 to 240 V, 50/60 Hz