missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKCSH Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£140.00 - £299.00
Specifications
| Antigen | PRKCSH |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18062594
|
Novus Biologicals
NBP3-16319-20UL |
20 μg |
£140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18311884
|
Novus Biologicals
NBP3-16319-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRKCSH Polyclonal antibody specifically detects PRKCSH in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PRKCSH | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AGE-R2,80K-H protein, G19P1AGE-binding receptor 2, glucosidase 2 subunit beta, Glucosidase II subunit beta, glucosidase II, beta subunit, hepatocystin, PCLD, PKCSH, PLD1, Protein kinase C substrate 60.1 kDa protein heavy chain, protein kinase C substrate 80K-H, protein kinase C substrate, 80 Kda protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-275 of human PRKCSH (P14314). | |
| Primary | |
| Store at 4°C. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 5589 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title