missing translation for 'onlineSavingsMsg'
Learn More

PRKCSH Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-16319-20UL

 View more versions of this product

Product Code. 18062594

  • £140.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PRKCSH Polyclonal antibody specifically detects PRKCSH in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRKCSH
Polyclonal
PBS, 50% glycerol, pH7.3
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
Unconjugated
AGE-R2,80K-H protein, G19P1AGE-binding receptor 2, glucosidase 2 subunit beta, Glucosidase II subunit beta, glucosidase II, beta subunit, hepatocystin, PCLD, PKCSH, PLD1, Protein kinase C substrate 60.1 kDa protein heavy chain, protein kinase C substrate 80K-H, protein kinase C substrate, 80 Kda protein
Recombinant fusion protein containing a sequence corresponding to amino acids 130-275 of human PRKCSH (P14314).
20 μg
Signal Transduction
5589
Store at 4°C.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.