missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKCSH Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-16319-100UL
This item is not returnable.
View return policy
Description
PRKCSH Polyclonal antibody specifically detects PRKCSH in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PRKCSH | |
| Polyclonal | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AGE-R2,80K-H protein, G19P1AGE-binding receptor 2, glucosidase 2 subunit beta, Glucosidase II subunit beta, glucosidase II, beta subunit, hepatocystin, PCLD, PKCSH, PLD1, Protein kinase C substrate 60.1 kDa protein heavy chain, protein kinase C substrate 80K-H, protein kinase C substrate, 80 Kda protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-275 of human PRKCSH (P14314). | |
| 100 μg | |
| Signal Transduction | |
| 5589 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction