Learn More
Potassium nitrate, ACS, 99.0% min, Thermo Scientific™
For Pyrotechnics, Glass Manufacture, Tempering Steel
£30.40 - £102.65
Chemical Identifiers
| CAS | 7757-79-1 |
|---|---|
| Molecular Formula | KNO3 |
| Molecular Weight (g/mol) | 101.102 |
| MDL Number | MFCD00011409 |
| InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
| Synonym | potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras |
| PubChem CID | 24434 |
| ChEBI | CHEBI:63043 |
| IUPAC Name | potassium;nitrate |
| SMILES | [N+](=O)([O-])[O-].[K+] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11366788
|
Thermo Scientific Alfa Aesar
013443.22 |
100g |
£30.40
100g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11356788
|
Thermo Scientific Alfa Aesar
013443.36 |
500g |
£51.40
500g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11376788
|
Thermo Scientific Alfa Aesar
013443.A3 |
2kg |
£114.00 £102.65 / 2kg Save £11.35 9% Off |
Estimated Shipment: 10-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| KNO3 | |
| MFCD00011409 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| CHEBI:63043 | |
| [N+](=O)([O-])[O-].[K+] |
Specifications
| 7757-79-1 | |
| KNO3 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| [N+](=O)([O-])[O-].[K+] | |
| 101.102 | |
| CHEBI:63043 | |
| ACS | |
| White crystalline | |
| Potassium nitrate, For ACS analysis |
| 99.0% min. | |
| MFCD00011409 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium;nitrate | |
| 24434 | |
| 101.11 | |
| Odorless | |
| 2.109g/cm3 |
Safety and Handling
H272-H303: May intensify fire; oxidizer. May be harmful if swallowed.
missing translation for 'dotInformation' : Hazard Class: 5.1, Packing Group: III
missing translation for 'einecsNumber' : 231-818-8
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.