missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, ACS, 99.0% min, Thermo Scientific™

For Pyrotechnics, Glass Manufacture, Tempering Steel

Brand:  Thermo Scientific Alfa Aesar 013443.36

Quantity 500g

 View more versions of this product

Product Code. 11356788

  • £51.40 / 500g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7757-79-1
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
potassium;nitrate
KNO3
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
CHEBI:63043
[N+](=O)([O-])[O-].[K+]

Specifications

7757-79-1
KNO3
UN1486
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
101.11
ACS
Hygroscopic
Potassium nitrate
500g
MFCD00011409
14,7648
Soluble in water,glycerol,liquid NH3Moderately soluble in water,and insoluble in alcohol; Soluble in glycerol and ammonia.
[N+](=O)([O-])[O-].[K+]
101.102
CHEBI:63043
≥99.0%
Odorless
2.109
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.