missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, ACS, 99.0% min

Product Code. 11376788
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376788 2 kg 2kg
11366788 100 g 100g
11356788 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11376788 Supplier Thermo Scientific Alfa Aesar Supplier No. 013443.A3
  • ONLINE EXCLUSIVE PRICE
    £114.00
    £102.65 / 2kg
    Save £11.35
    9% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Pyrotechnics, Glass Manufacture, Tempering Steel | CAS: 7757-79-1 | KNO3 | 101.10 g/mol

Pyrotechnics, glass manufacture, tempering steel, reagentPotassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O

Specifications

Boiling Point 400°C (decomposition)
Melting Point 334°C
Color White
Physical Form Crystals
Quantity 2 kg
Assay Percent Range 99.0% min.
Solubility Information Soluble in water,glycerol,liquid NH3Moderately soluble in water,and insoluble in alcohol. Soluble in glycerol and ammonia.
IUPAC Name potassium nitrate
Formula Weight 101.11
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Density 2.109
Chemical Name or Material Potassium nitrate, For ACS analysis
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.