missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PMF1 Antibody (SR4288), Novus Biologicals™

Rabbit Monoclonal Antibody

£240.00 - £290.00

Specifications

Antigen PMF1
Clone SR4288
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30041627
View Documents
Novus Biologicals™
NBP3-27835-100ul
100 μL
£290.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30041746
View Documents
Novus Biologicals™
NBP3-27835-50ul
50 μL
£240.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PMF1 Monoclonal antibody specifically detects PMF1 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications

Specifications

PMF1
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human, Mouse, Rat
PMF-1, Polyamine Modulated Factor 1
A synthesized peptide derived from human PMF1 (Uniprot #: Q6P1K2)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
SR4288
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Cell Biology, Cell Cycle and Replication
PBS (pH 7.4), 150mM NaCl, 50% glycerol
11243
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.