missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PMF1 Antibody (SR4288), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27835-50ul
This item is not returnable.
View return policy
Description
PMF1 Monoclonal antibody specifically detects PMF1 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PMF1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunoprecipitation, Western Blot | |
| SR4288 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| PMF-1, Polyamine Modulated Factor 1 | |
| A synthesized peptide derived from human PMF1 (Uniprot #: Q6P1K2) | |
| 50 μL | |
| Cell Biology, Cell Cycle and Replication | |
| 11243 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction