missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PMF1 Antibody (SR4288), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-27835-100ul

 View more versions of this product

Product Code. 30041627

  • £290.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PMF1 Monoclonal antibody specifically detects PMF1 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PMF1
Monoclonal
Unconjugated
PBS (pH 7.4), 150mM NaCl, 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Immunoprecipitation, Western Blot
SR4288
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
PMF-1, Polyamine Modulated Factor 1
A synthesized peptide derived from human PMF1 (Uniprot #: Q6P1K2)
100 μL
Cell Biology, Cell Cycle and Replication
11243
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.