Learn More
PeproTech PlGF-1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
£177.00 - £2745.00
Specifications
| Antigen | PlGF-1 |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17855213
![]() |
PeproTech
500-P226BT-25UG |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17884923
![]() |
PeproTech
500-P226BT-50UG |
50 μg |
£228.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17875273
|
Invitrogen
500-P226BT-1MG |
2 x 500 μg | |||||||
|
30054669
|
PeproTech
500-P226BT-500UG |
500 μg |
£1466.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: MLPAVPPQQW ALSAGNGSSE VEVVPFQEVW GRSYCRALER LVDVVSEYPS EVEHMFSPSC VSLLRCTGCC GDENLHCVPV ETANVTMQLL KIRSGDRPSY VELTFSQHVR CECRPLREKM KPERCGDAVP RR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PlGF-1. Anti-Human PlGF-1-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hPlGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human PlGF (500-P226) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hPlGF. Western Blot: To detect hPlGF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hPlGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P226BT-1MG will be provided as 2 x 500 μg
PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, the colon, and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration.Specifications
| PlGF-1 | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Human | |
| P49763 | |
| PGF | |
| Primary | |
| -20°C |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| 5228 | |
| E.coli-derived, 29.7 kDa Recombinant Human PlGF-1. | |
| Antigen affinity chromatography | |
| PGF |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.