missing translation for 'onlineSavingsMsg'
Learn More

PeproTech PlGF-1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P226BT-500UG

 View more versions of this product

Product Code. 30054669

  • £1466.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: MLPAVPPQQW ALSAGNGSSE VEVVPFQEVW GRSYCRALER LVDVVSEYPS EVEHMFSPSC VSLLRCTGCC GDENLHCVPV ETANVTMQLL KIRSGDRPSY VELTFSQHVR CECRPLREKM KPERCGDAVP RR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PlGF-1. Anti-Human PlGF-1-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hPlGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human PlGF (500-P226) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hPlGF. Western Blot: To detect hPlGF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hPlGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P226BT-1MG will be provided as 2 x 500 μg

PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, the colon, and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PlGF-1
Polyclonal
Biotin
PGF
Placenta Growth Factor; Placenta Growth Factor-1
Rabbit
Antigen affinity chromatography
RUO
5228
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P49763
PGF
E.coli-derived, 29.7 kDa Recombinant Human PlGF-1.
500 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.