Learn More
Platinum 13 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type R Standard Grade Thermocouple, Thermo Scientific Chemicals
For ISA Type R Standard Grade Thermocouple
POA - £1916.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11342118
|
Thermo Scientific Alfa Aesar
013018.BY |
100 cm |
£1916.00
100cm |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11332118
|
Thermo Scientific Alfa Aesar
013018.BU |
25 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Platinum 13 wt% Rhodium wire, 0.5mm (0.02in) dia is used as thermocouples and heat flux gauges. It can be used for high temperature measurements because high cost and low sensitivity.