Learn More
Platinum 13 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type R Standard Grade Thermocouple, Thermo Scientific Chemicals
For ISA Type R Standard Grade Thermocouple
Brand: Thermo Scientific Alfa Aesar 013018.BU
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Platinum 13 wt% Rhodium wire, 0.5mm (0.02in) dia is used as thermocouples and heat flux gauges. It can be used for high temperature measurements because high cost and low sensitivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum 13 wt% Rhodium wire | |
Pt:Rh; 87:13 wt% | |
MFCD00198186 | |
Insoluble in water. | |
19.595g/cc |
0.5mm (0.02 in.) dia., Positive leg Type R | |
19.595G/CC | |
25 cm | |
Odorless | |
Wire |
RUO – Research Use Only