missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SM(PEG)12 (PEGylated, long-chain SMCC crosslinker)

Product Code. 10117443 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10117443 100 mg 100mg
10381444 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10117443 Supplier Thermo Scientific™ Supplier No. 22112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Crosslink amines to sulfhydryls with these crosslinkers containing NHS-ester and maleimide groups at ends of water-soluble polyethylene glycol spacer arms (17.6 to 95.2A).

Thermo Scientific™ Pierce SM(PEG)n is a series of amine-to-sulfhydryl crosslinkers that differ in length from 17.6 to 95.2 Ås as a result of polyethylene glycol spacer arms containing n equals 2 to 24 ethylene glycol units.

The SM(PEG)n reagents are six crosslinkers that have identical amine- and sulfhydryl-reactive groups but differ in length as a result of spacer arms composed of different numbers of ethylene glycol units. Their structures (shown below) have the form NHS-(PEG)n-Maleimide, where n represents 2, 4, 6, 8, 12 or 24 polyethylene glycol (PEG) units. Each SM(PEG)n reagent is produced at the specified length without contamination from analogs of other lengths.The availability of this set of reagents makes it possible to optimize crosslinking experiments to obtain the best conjugate efficiency and function for the intended application or analysis method. Considered together with AMAS (Part No. 22295) and the popular SMCC (Part No. 22360), these reagents allow for amine-to-sulfhydryl crosslinking across spaces ranging from 4.4 to 95.2Å. In addition, the hydrophilic PEG content of SM(PEG)n reagents adds to the aqueous solubility of these crosslinkers and their conjugates. PEG is also less immunogenic than other types of crosslinker spacers, making these reagents ideal for immunogen preparation.

Highlights:

  • Homologous series of increasing spacer arm length, ideal for optimizing conjugation of protein interactions
  • Water-soluble polyethylene glycol (PEG) spacer arms help to maintain or increase solubility of crosslinked proteins
  • Reagents readily dissolve in DMSO for subsequent dilution in aqueous reaction buffers
  • Reactive groups: NHS ester and maleimide
  • Reactive toward: amino and sulfhydryl groups

NHS-PEG2-Maleimide

  • Two ethylene oxide subunits
  • Alternate names: Mal-dPEG2-NHS, NHS-PEG2-Maleimide
  • Molecular formula C18H23N3O9
  • Spacer arm length 17.6Å
  • Mass added to target per crosslink 310.12
  • MW 425.39

NHS-PEG4-Maleimide

  • Four ethylene oxide subunits
  • Alternate names Mal-dPEG4-NHS, NHS-PEG4-Maleimide
  • Molecular formula C22H31N3O11
  • Spacer arm length 24.6Å
  • Mass added to target per crosslink 398.17
  • MW 513.50&

NHS-PEG6-Maleimide

  • Six ethylene oxide subunits
  • Alternate names Mal-dPEG6-NHS, NHS-PEG6-Maleimide
  • Molecular formula C26H39N3O13
  • Spacer arm length 32.25Å
  • Mass added to target per crosslink 486.20
  • MW 601.60

NHS-PEG8-Maleimide

  • Eight ethylene oxide subunits
  • Alternate names Mal-dPEG8-NHS, NHS-PEG8-Maleimide
  • Molecular formula C30H47N3O15
  • Spacer arm length 39.2Å
  • Mass added to target per crosslink 574.27
  • MW 689.71

NHS-PEG12-Maleimide

  • 12 ethylene oxide subunits Mal-dPEG12-NHS, NHS-PEG12-Maleimide
  • Molecular Formula C38H63N3O19
  • Spacer arm length 53.4Å
  • Mass added to target per crosslink 750.38
  • MW 865.92
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable No
Molecular Weight (g/mol) 865.92
PEGylated Yes
Spacer Arm Length 53.4 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, NHS Ester
Spacer Long (>30 Å)
Form Liquid
Quantity 100 mg
Solubility Water
Format Standard, Single-use
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.