Learn More
Thermo Scientific™ SBAP (succinimidyl 3-(bromoacetamido)propionate)
Crosslink amines to sulfhydryls with this short crosslinker composed of NHS-ester and bromoacetyl (haloacetyl) groups at ends of a 7-atom (6.2A) spacer arm.
Brand: Thermo Scientific™ 22339
Code : PN
Additional Details : CAS Number : 2899-30-1 Weight : 0.05000kg
Description
Thermo Scientific™ Pierce SBAP is a short (6.2 Å) crosslinker for amine-to-sulfhydryl conjugation via N-hydroxysuccinimide (NHS) ester and bromoacetyl reactive groups.
Highlights:
- Useful for preparing cyclic peptides and peptide conjugates
- Reactive groups: NHS ester and bromoacetyl
- Reactive toward: amino and sulfhydryl groups
Specifications
Amine-Sulfhydryl | |
SBAP | |
No | |
Upon receipt store at 4°C. | |
Ambient | |
Chemical Labeling | |
Haloacetyl, NHS Ester | |
Solid | |
DMF, DMSO | |
No |
No | |
307.1 | |
6.2 Å | |
Yes | |
Pierce™ | |
Heterobifunctional | |
Short (<10 Å) | |
50 mg | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.