Learn More
Thermo Scientific™ L-Photo-Methionine
Shop All Thermo Scientific ProductsDescripción
Thermo Scientific™ Pierce Photoreactive Amino Acids are analogs of L-Leucine and L-Methionine that can be incorporated into proteins during synthesis and then light-activated to covalently crosslink interacting proteins in cells.
Highlights:
In vivo labeling – incorporate photo-reactive group into proteins using normal cellular machinery
In vivo crosslinking – find interacting proteins in the native cellular environment
Increased specificity compared to traditional methods – crosslink interacting proteins correctly positioned at their interfaces within protein interaction domains
Complementary to traditional methods – provides detailed characterization of protein interactions when compared to results with formaldehyde and other more general crosslinking approaches
Efficient recovery – 90% protein recovery in cell lysates after crosslinking
Compatible – crosslink proteins expressed in a wide variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3 and C6
Easy to use – reagents are photo-stable under normal laboratory lighting, eliminating the need to work in the dark
Especificaciones
Especificaciones
| Chemical Reactivity | Nonselective-NA (Metabolic labeling) |
| Cleavable | No |
| Description | L-Photo-Methionine |
| Molecular Weight (g/mol) | 157.17 |
| PEGylated | No |
| Spacer Arm Length | 0.0 Å |
| Content And Storage | Upon receipt store at 4°C protected from light. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Mostrar más |
For Research Use Only. Not for use in diagnostic procedures.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.