Learn More
Thermo Scientific™ NHS-Phosphine
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce NHS-Phosphine and Sulfo-NHS-Phosphine are amine-reactive compounds that can be used to derivatize primary amines of proteins or amine-coated polymer surfaces for ligation to azide-tagged molecules.
Highlights:
Versatile – Sulfo-NHS-Phosphine easily dissolves in water-miscible solvents for subsequent dilution in aqueous reaction mixtures for azide conjugation; NHS-Phosphine can be used for amine surface modification in organic solvents
Compatible – reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents
Specific – NHS esters are specific for covalent attachment to primary amines (side chain of lysine or N-terminus of polypeptides)
Chemoselective – azide and phosphine groups do not react or interfere with components of biological samples but conjugate to one another with high efficiency
Specifications
Specifications
| Chemical Reactivity | Amine-Azide |
| Cleavable | No |
| Description | NHS-Phosphine |
| Molecular Weight (g/mol) | 461.4 |
| PEGylated | No |
| Spacer Arm Length | 5.4 Å |
| Content And Storage | Upon receipt store at -20°C protected from light and moisture. |
| Cell Permeability | Not determined |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.