Learn More
Thermo Scientific™ SDA (NHS-Diazirine) (succinimidyl 4,4'-azipentanoate)
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce NHS-Diazirine Crosslinkers combine proven NHS-ester and diazirine-based photoreaction chemistries to conjugate amine-containing molecules with nearly any other functional group via long-wave UV-light activation.
Highlights:
Versatile - different amine-reactive diazirine derivatives capture specific intracellular or cell-surface protein interactions
Controllable - two-step chemical crosslinking is activated using common laboratory UV lamps
Reversible - disulfide-containing spacers enable cleavage using reducing agents
Easy - reagents are photo-stable under typical laboratory lighting conditions so there is no need to perform experiments in the dark
Better than aryl azides - diazirine photo-reactive group has better photostability and is more efficiently activated by long-wave UV light
Specifications
Specifications
| Chemical Reactivity | Amine-Nonselective |
| Cleavable | No |
| Description | SDA |
| Molecular Weight (g/mol) | 225.2 |
| PEGylated | No |
| Spacer Arm Length | 3.9 Å |
| Content And Storage | Upon receipt store product at 4°C protected from light and moisture. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.