Learn More
Description
Thermo Scientific™ Pierce NHS-Azide reagents are amine-reactive compounds that can be used to derivatize primary amines of proteins or amine-coated polymer surfaces for ligation to phosphine-modified molecules.
These NHS-ester compounds react to form covalent bonds with primary amines (e.g., side chain of lysine residues or aminosilane-coated surfaces). The azide (N3) group reacts with phosphine-labeled molecules by a mechanism known as Staudinger chemistry, enabling efficient and specific conjugation of derivatized molecules in biological samples. In combination with different phosphine-activated compounds, these NHS-Azide reagents facilitate a variety of labeling and crosslinking experimental strategies for investigating protein interactions and cellular pathways.
Highlights:
Soluble – reagents easily dissolve in water-miscible solvents for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples
Compatible – reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents
Specific – NHS esters are specific for covalent attachment to primary amines (side chain of lysine or N-terminus of polypeptides)
Chemoselective – azide and phosphine groups do not react or interfere with components of biological samples but conjugate to one another with high efficiency
Pegylated – choose the very short NHS-Azide or one of the two varieties whose spacer arms contain medium (4-unit) or long (12-unit) polyethylene glycol (PEG) groups for increased solubility and reach
Specifications
Specifications
| Chemical Reactivity | Amine-Phosphine |
| Cleavable | No |
| Description | NHS-Azide |
| Molecular Weight (g/mol) | 198.14 |
| PEGylated | No |
| Spacer Arm Length | 2.5 Å |
| Content And Storage | Upon receipt store at -20°C protected from light and moisture. |
| Cell Permeability | Not determined |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.