Learn More
Thermo Scientific™ MPBH (4-(4-N-maleimidophenyl)butyric acid hydrazide)
Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this long (17.9A) crosslinker containing maleimide and hydrazide groups.
Brand: Thermo Scientific™ 22305
Code : PN
Additional Details : CAS Number : 66085-59-4 Weight : 0.80000kg
Description
Thermo Scientific™ Pierce MPBH is a long, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).
Highlights:
- Sulfhydryl-reactive maleimide forms a thioether linkage
- Stable in DMSO
- Reactive groups: hydrazide and maleimide
- Reactive toward: carbohydrate and sulfhydryl groups
- Use with EDC for reactivity toward carboxyl groups
Specifications
Carbohydrate-Sulfhydryl | |
MPBH | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
Amine, Hydrazide, Maleimide | |
Powder | |
DMF, DMSO | |
No |
No | |
309.75 | |
17.9 Å | |
Yes | |
Pierce™ | |
Heterobifunctional | |
Medium (10 to 30 Å) | |
50 mg | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.