Learn More
Thermo Scientific™ DSS (disuccinimidyl suberate), No-Weigh™ Format
Crosslink amines to amines with disuccinimidyl suberate composed of NHS-ester groups with a membrane-permeable, mid-length spacer for intracellular protein crosslinking.
Brand: Thermo Scientific™ 21555
Code : PN
Additional Details : CAS Number : 68528-80-3 Weight : 1.81440kg
Description
Thermo Scientific™ Pierce DSS, also called disuccinimidyl suberate, is noncleavable and membrane permeable crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm.
Highlights:
- Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
- “Fix” protein interactions to allow identification of weak or transient protein interactions
- Protein crosslinking to create bioconjugates via single-step reactions
- Crosslink oriented antibodies to Protein A or G agarose
- Immobilize proteins onto amine-coated surfaces
Specifications
Amine-Amine | |
DSS | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
NHS Ester | |
Powder | |
DMF, DMSO | |
No |
No | |
368.35 | |
11.4 Å | |
Yes | |
Pierce™ | |
Homobifunctional | |
Medium (10 to 30 Å) | |
1 g | |
Standard, Single-use | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.