missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DMS (dimethyl suberimidate)

Product Code. 10638674 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10638674

Brand: Thermo Scientific™ 20700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Crosslink amines to amines with dimethyl suberimidate (CAS 34490-86-3); composed of imidoester groups at either end of a membrane-permeable, 8-atom spacer arm.

Thermo Scientific™ Pierce DMS is dimethyl suberimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of an 8-atom spacer arm.

Highlights:

  • Water-soluble
  • Rapid reaction with amines at alkaline pH values (pH 8-10)
  • Amidine bond retains net charge character of protein
  • Reversible at high pH values
  • Reactive groups: imidoester (homobifunctional)
  • Reactive toward: amino groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description DMS
Molecular Weight (g/mol) 273.2
PEGylated No
Spacer Arm Length 11.0 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Imidoester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 1 g
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.