missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BMB (1,4-bismaleimidobutane)

Product Code. 10046663 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10046663 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10046663 Supplier Thermo Scientific™ Supplier No. 22331

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Crosslink sulfhydryls to sulfhydryls (cysteine residues in proteins or peptides) with this mid-length crosslinker composed of maleimide groups and 9-atom (10.9A) spacer arm.

Thermo Scientific™ Pierce BMB is a mid-length, maleimide crosslinker for covalent, irreversible conjugation between sulfhydryl groups (e.g., protein or peptide cysteines).

Highlights:

  • Reactive groups: maleimide (homobifunctional)
  • Reactive toward: sulfhydryl groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Sulfhydryl-Sulfhydryl
Cleavable No
Description BMB
Molecular Weight (g/mol) 248.23
PEGylated No
Spacer Arm Length 10.9 Å
Content And Storage Upon receipt store at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Maleimide
Spacer Medium (10 to 30 Å)
Form Solid
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
compliance-icons
Product Identifier
  • BMB (1,4-bismaleimidobutane)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-BisMaleimidobutane

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.