missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase A2 XII Antibody (7C7C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£187.00 - £435.00
Specifications
| Antigen | Phospholipase A2 XII |
|---|---|
| Clone | 7C7C9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18689525
|
Novus Biologicals
NBP2-37258-0.025ml |
0.025 mL |
£187.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18116447
|
Novus Biologicals
NBP2-37258 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospholipase A2 XII Monoclonal specifically detects Phospholipase A2 XII in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Phospholipase A2 XII | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 3.1.1.4, group XII secreted phospholipase A2, group XIIA secreted phospholipase A2, group XIIA secretory phospholipase A2, GXII, GXII sPLA2, Phosphatidylcholine 2-acylhydrolase 12A, phospholipase A2, group XII, phospholipase A2, group XIIA, PLA2G12, ROSSY, sPLA2-XII | |
| PLA2G12A | |
| IgG1 | |
| Affinity Purified |
| 7C7C9 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Lipid and Metabolism | |
| PBS with 0.05% Sodium Azide | |
| 81579 | |
| Purified recombinant fragment of human PLA2G12A (AA: 21-189) expressed in E. Coli. | |
| Primary | |
| 21 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title