missing translation for 'onlineSavingsMsg'
Learn More

Phospholipase A2 XII Antibody (7C7C9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-37258

 View more versions of this product

Product Code. 18116447

  • £435.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Phospholipase A2 XII Monoclonal specifically detects Phospholipase A2 XII in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospholipase A2 XII
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready
EC 3.1.1.4, group XII secreted phospholipase A2, group XIIA secreted phospholipase A2, group XIIA secretory phospholipase A2, GXII, GXII sPLA2, Phosphatidylcholine 2-acylhydrolase 12A, phospholipase A2, group XII, phospholipase A2, group XIIA, PLA2G12, ROSSY, sPLA2-XII
Mouse
21 kDa
0.1 mL
Lipid and Metabolism
81579
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Flow Cytometry, ELISA, CyTOF
7C7C9
Unconjugated
PBS with 0.05% Sodium Azide
PLA2G12A
Purified recombinant fragment of human PLA2G12A (AA: 21-189) expressed in E. Coli.
Affinity Purified
RUO
Primary
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only