missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase A2 XII Antibody (7C7C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37258-0.025ml
This item is not returnable.
View return policy
Description
Phospholipase A2 XII Monoclonal specifically detects Phospholipase A2 XII in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| Phospholipase A2 XII | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| EC 3.1.1.4, group XII secreted phospholipase A2, group XIIA secreted phospholipase A2, group XIIA secretory phospholipase A2, GXII, GXII sPLA2, Phosphatidylcholine 2-acylhydrolase 12A, phospholipase A2, group XII, phospholipase A2, group XIIA, PLA2G12, ROSSY, sPLA2-XII | |
| Mouse | |
| 21 kDa | |
| 0.025 mL | |
| Lipid and Metabolism | |
| 81579 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 7C7C9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PLA2G12A | |
| Purified recombinant fragment of human PLA2G12A (AA: 21-189) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction