missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGLYRP3/PGRPIA Antibody (187C434), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
£167.00 - £302.00
Specifications
| Antigen | PGLYRP3/PGRPIA |
|---|---|
| Clone | 187C434 |
| Dilution | Western Blot, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Uehara et al (2005)), Flow (Cell Surface) reported in scientific literature (PMID 15839897), Flow (Intracellular) reported in scientific literature (PMID 16849490) |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18068255
|
Novus Biologicals
NB100-56729SS |
0.05 mL |
£167.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18085645
|
Novus Biologicals
NB100-56729 |
0.2 mL |
£302.00
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGLYRP3/PGRPIA Monoclonal specifically detects PGLYRP3/PGRPIA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), Flow (Intracellular).Specifications
| PGLYRP3/PGRPIA | |
| Western Blot, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Uehara et al (2005)), Flow (Cell Surface) reported in scientific literature (PMID 15839897), Flow (Intracellular) reported in scientific literature (PMID 16849490) | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.05% Sodium Azide | |
| peptidoglycan recognition protein 3, peptidoglycan recognition protein I alpha, Peptidoglycan recognition protein I-alpha, Peptidoglycan recognition protein intermediate alpha, peptidoglycan recognition protein-I-alpha, PGLYRPIalphaMGC149197, PGRP-Ialpha, PGRPIAPGRP-I-alpha | |
| PGLYRP3 | |
| IgM | |
| Unpurified | |
| PGRP-1a (187C434) |
| 187C434 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96LB9 | |
| 114771 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 178-193 of human PGRP-1a. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title