missing translation for 'onlineSavingsMsg'
Learn More

PGLYRP3/PGRPIA Antibody (187C434), Novus Biologicals™

Product Code. 18068255 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
Unit Size:
0.05mL
0.2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18068255 0.05 mL 0.05mL
18085645 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18068255 Supplier Novus Biologicals Supplier No. NB10056729SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

PGLYRP3/PGRPIA Monoclonal specifically detects PGLYRP3/PGRPIA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), Flow (Intracellular).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGLYRP3/PGRPIA
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 187C434
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Uehara et al (2005)), Flow (Cell Surface) reported in scientific literature (PMID 15839897), Flow (Intracellular) reported in scientific literature (PMID 16849490)
Formulation Ascites with 0.05% Sodium Azide
Gene Accession No. Q96LB9
Gene Alias peptidoglycan recognition protein 3, peptidoglycan recognition protein I alpha, Peptidoglycan recognition protein I-alpha, Peptidoglycan recognition protein intermediate alpha, peptidoglycan recognition protein-I-alpha, PGLYRPIalphaMGC149197, PGRP-Ialpha, PGRPIAPGRP-I-alpha
Gene Symbols PGLYRP3
Host Species Mouse
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 178-193 of human PGRP-1a.
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114771
Test Specificity PGRP-1a (187C434)
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.