missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGLYRP3/PGRPIA Antibody (187C434), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB100-56729
This item is not returnable.
View return policy
Description
PGLYRP3/PGRPIA Monoclonal specifically detects PGLYRP3/PGRPIA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), Flow (Intracellular).
Specifications
| PGLYRP3/PGRPIA | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.05% Sodium Azide | |
| peptidoglycan recognition protein 3, peptidoglycan recognition protein I alpha, Peptidoglycan recognition protein I-alpha, Peptidoglycan recognition protein intermediate alpha, peptidoglycan recognition protein-I-alpha, PGLYRPIalphaMGC149197, PGRP-Ialpha, PGRPIAPGRP-I-alpha | |
| Mouse | |
| Unpurified | |
| RUO | |
| 114771 | |
| Human | |
| Ascites |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 187C434 | |
| Western Blot, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Uehara et al (2005)), Flow (Cell Surface) reported in scientific literature (PMID 15839897), Flow (Intracellular) reported in scientific literature (PMID 16849490) | |
| Q96LB9 | |
| PGLYRP3 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 178-193 of human PGRP-1a. | |
| 0.2 mL | |
| Primary | |
| PGRP-1a (187C434) | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction