missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE5A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£201.00 - £559.00
Specifications
| Antigen | PDE5A |
|---|---|
| Dilution | Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:250 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18234671
|
Novus Biologicals
NB300-655-0.025MG |
0.025 mg |
£201.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18276622
|
Novus Biologicals
NB300-655-0.05MG |
0.05 mg |
£341.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18179049
|
Novus Biologicals
NB300-655 |
0.1 mg |
£559.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDE5A Polyclonal specifically detects PDE5A in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| PDE5A | |
| Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| 8654 | |
| Synthetic cyclic peptide (common to all PDE5A variants) (Rat). | |
| Primary |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:250 | |
| Polyclonal | |
| Rabbit | |
| PDE5A | |
| IgG | |
| This detects 99 and 89 kDa PDE5A proteins in various tissues. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title