missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE5A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-655-0.05MG
This item is not returnable.
View return policy
Description
PDE5A Polyclonal specifically detects PDE5A in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| PDE5A | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide | |
| PDE5A | |
| Synthetic cyclic peptide (common to all PDE5A variants) (Rat). | |
| Primary | |
| This detects 99 and 89 kDa PDE5A proteins in various tissues. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.50 mg/mL | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:250 | |
| CGB-PDE, cGMP-binding cGMP-specific 3′-5′-cyclic nucleotide phosphodiesterase, cGMP-binding cGMP-specific phosphodiesterase, cGMP-specific 3′-5′-cyclic phosphodiesterase, cGMP-specific phosphodiesterase type 5A, CN5A, EC 3.1.4, EC 3.1.4.35, PDE5, phosphodiesterase 5A, cGMP-specific, phosphodiesterase isozyme 5 | |
| Rabbit | |
| 0.05 mg | |
| 8654 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction