missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE5A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-655
This item is not returnable.
View return policy
Description
PDE5A Polyclonal antibody specifically detects PDE5A in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Specifications
| PDE5A | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| PDE5A | |
| Synthetic cyclic peptide (common to all PDE5A variants) (Rat). | |
| 0.1 mg | |
| Lipid and Metabolism, Signal Transduction | |
| 8654 | |
| Human, Mouse, Rat, Chicken | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.5 mg/mL | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:250 | |
| CGB-PDE, cGMP-binding cGMP-specific 3'-5'-cyclic nucleotide phosphodiesterase, cGMP-binding cGMP-specific phosphodiesterase, cGMP-specific 3'-5'-cyclic phosphodiesterase, cGMP-specific phosphodiesterase type 5A, CN5A, EC 3.1.4, EC 3.1.4.35, PDE5, phosphodiesterase 5A, cGMP-specific, phosphodiesterase isozyme 5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This detects 99 and 89 kDa PDE5A proteins in various tissues. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction