missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Paxillin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Provide accurate, reproducible results with the Epredia™ Paxillin Ab-1, Mouse Monoclonal Antibody.
£120.36 - £601.80
Specifications
Antigen | Paxillin Ab-1 |
---|---|
Clone | 5H11 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12611357
|
Epredia™
MS-404-P1 |
Concentrated | 500 μL |
£385.00
500µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12621357
|
Epredia™
MS-404-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12631357
|
Epredia™
MS-404-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12601357
|
Epredia™
MS-404-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12681347
|
Epredia™
MS-404-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12671357
|
Epredia™
MS-404-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.Host Species: Mouse
Clone: 5H11
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant full-length human paxillin protein
Molecular Weight: 68kDa
Positive Control: HeLa cells, Colon CA
Cellular Localization: Membrane and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Paxillin Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
IgG2a κ |
5H11 | |
Monoclonal | |
Mouse | |
Cytoskeletal Protein | |
Purified recombinant full-length human paxillin protein | |
Primary |