missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p57 Kip2 Ab-3, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Deliver accurate, reproducible results with the Epredia™ p57Kip2 Ab-3, Mouse Monoclonal Antibody.
£99.91 - £576.30
Specifications
Antigen | p57superscriptKip2superscript |
---|---|
Clone | KP39 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Monoclonal |
Form | Concentrated |
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
12652557
|
Epredia
MS-897-P0 |
Concentrated | 200μg/mL | 100 μL | |||||
12622557
|
Epredia
MS-897-B1 |
Concentrated | 200μg/mL | 500 μL | |||||
12672557
|
Epredia
MS-897-P1 |
Concentrated | 200μg/mL | 500 μL |
£339.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12612557
|
Epredia
MS-897-B |
Concentrated | 200μg/mL | 1 mL | |||||
12642557
|
Epredia
MS-897-P |
Concentrated | 200μg/mL | 1 mL | |||||
12692557
|
Epredia
MS-897-P1ABX |
Concentrated | 1mg/mL | 100 μL | |||||
12682557
|
Epredia
MS-897-PABX |
Concentrated | 1mg/mL | 200 μL | |||||
Description
p57Kip2 (or CDKN1C) is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. The gene encoding human p57Kip2 is located on chromosome 11p15.5, a region implicated in both sporadic cancers, Wilm's tumor, and Beckwith-Wiedemann syndrome (BWS), a cancer syndrome, making it a tumor suppressor candidate. BWS is characterized by numerous growth abnormalities and an increased risk of childhood tumors. Several types of childhood tumors including Wilms' tumor, adrenocortical carcinoma and rhabdomyosarcoma display a specific loss of maternal 11p15 alleles, suggesting that genomic imprinting plays an important part. This region also contains two other imprinted genes, insulin-like growth factor II (IGF-II) and H19, both of which seem to be implicated in adrenal neoplasms.Host Species: Mouse
Clone: KP39
Isotype: IgG2b/κ
Species Reactivity: Human and Mouse. Others not known.
Epitope: Not determined
Immunogen: Recombinant human p57Kip2 protein
Molecular Weight: 57kDa
Positive Control: LS174T cells, Colon CA or placenta
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not suitable)
- Immunoprecipitation (Native verified)
Specifications
p57superscriptKip2superscript | |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
Concentrated | |
IVD | |
Human, Mouse | |
IgG2b κ |
KP39 | |
Monoclonal | |
Mouse | |
Cell Cycle | |
Recombinant human p57superscriptKip2superscript protein | |
Primary |