Learn More
p57 Kip2 Ab-3, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Deliver accurate, reproducible results with the Epredia™ p57Kip2 Ab-3, Mouse Monoclonal Antibody.
Brand: Epredia MS-897-P1
Description
p57Kip2 (or CDKN1C) is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. The gene encoding human p57Kip2 is located on chromosome 11p15.5, a region implicated in both sporadic cancers, Wilm's tumor, and Beckwith-Wiedemann syndrome (BWS), a cancer syndrome, making it a tumor suppressor candidate. BWS is characterized by numerous growth abnormalities and an increased risk of childhood tumors. Several types of childhood tumors including Wilms' tumor, adrenocortical carcinoma and rhabdomyosarcoma display a specific loss of maternal 11p15 alleles, suggesting that genomic imprinting plays an important part. This region also contains two other imprinted genes, insulin-like growth factor II (IGF-II) and H19, both of which seem to be implicated in adrenal neoplasms.Host Species: Mouse
Clone: KP39
Isotype: IgG2b/κ
Species Reactivity: Human and Mouse. Others not known.
Epitope: Not determined
Immunogen: Recombinant human p57Kip2 protein
Molecular Weight: 57kDa
Positive Control: LS174T cells, Colon CA or placenta
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not suitable)
- Immunoprecipitation (Native verified)
Specifications
| p57superscriptKip2superscript | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Cell Cycle | |
| Human, Mouse | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| KP39 | |
| Unconjugated | |
| Recombinant human p57superscriptKip2superscript protein | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.