missing translation for 'onlineSavingsMsg'
Learn More

OXSR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen OXSR1
Concentration 0.13 mg/mL
Applications Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16894854
View Documents
Proteintech
15611-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16884854
View Documents
Proteintech
15611-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Regulates downstream kinases in response to environmental stress. May also have a function in regulating the actin cytoskeleton.
Specifications

Specifications

OXSR1
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
O95747, Q6P9R2
108737, 316064, 9943
OXSR1 Fusion Protein Ag8002
Primary
-20°C
OXSR1
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
KIAA1101, OSR1, oxidative stress responsive 1, OXSR1
Oxsr1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.