missing translation for 'onlineSavingsMsg'
Learn More

OXSR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16884854 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16884854 150 μL 150µL
16894854 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16884854 Supplier Proteintech Supplier No. 156111AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Regulates downstream kinases in response to environmental stress. May also have a function in regulating the actin cytoskeleton.

Specifications

Antigen OXSR1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OXSR1
Gene Accession No. O95747, Q6P9R2
Gene Alias KIAA1101, OSR1, oxidative stress responsive 1, OXSR1
Gene Symbols Oxsr1
Host Species Rabbit
Immunogen OXSR1 Fusion Protein Ag8002
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108737, 316064, 9943
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.