missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Regulates downstream kinases in response to environmental stress. May also have a function in regulating the actin cytoskeleton.
Specifications
Specifications
| Antigen | OXSR1 |
| Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | OXSR1 |
| Gene Accession No. | O95747, Q6P9R2 |
| Gene Alias | KIAA1101, OSR1, oxidative stress responsive 1, OXSR1 |
| Gene Symbols | Oxsr1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?