missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXPAT Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£186.00 - £417.00
Specifications
| Antigen | OXPAT |
|---|---|
| Concentration | 1.01 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Simple Western 10 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18137069
|
Novus Biologicals
NB110-60511SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18781114
|
Novus Biologicals
NB110-60511 |
£417.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
OXPAT Polyclonal specifically detects OXPAT in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
| OXPAT | |
| Western Blot 0.5-2 ug/ml, Simple Western 10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| 440503 | |
| A synthetic peptide made to a C-terminal region of the mouse OXPAT protein, within residues 400-463. [UniProt# Q8BVZ1] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 50 kDa |
| 1.01 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| lipid storage droplet protein 5, LSDA5, LSDP5, MLDP, OXPAT, perilipin 5, perilipin-5 | |
| PLIN5 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Immunogen sequence has 89% homology to rat and 75% homology to bovine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title