Learn More
OXPAT Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB110-60511
This item is not returnable.
View return policy
Description
OXPAT Polyclonal specifically detects OXPAT in Human, Mouse samples. It is validated for Western Blot, Simple Western.
Specifications
| Western Blot 0.5-2 ug/ml, Simple Western 10 ug/ml | |
| lipid storage droplet protein 5, LSDA5, LSDP5, MLDP, OXPAT, perilipin 5, perilipin-5 | |
| PLIN5 | |
| Affinity Purified |