missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXPAT Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB110-60511SS
This item is not returnable.
View return policy
Description
OXPAT Polyclonal specifically detects OXPAT in Human, Mouse samples. It is validated for Western Blot, Simple Western.
Specifications
| OXPAT | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| PLIN5 | |
| A synthetic peptide made to a C-terminal region of the mouse OXPAT protein, within residues 400-463. [UniProt# Q8BVZ1] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen sequence has 89% homology to rat and 75% homology to bovine. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.01 mg/mL | |
| Western Blot 0.5-2 ug/ml, Simple Western 10 ug/ml | |
| lipid storage droplet protein 5, LSDA5, LSDP5, MLDP, OXPAT, perilipin 5, perilipin-5 | |
| Rabbit | |
| 50 kDa | |
| 0.025 mL | |
| Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
| 440503 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction